Learn More
growth differentiation factor 11, Mouse, Clone: 3G6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GDF11.
Marca: Abnova H00010220-M01.100ug
Descrizione
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice and Xenopus suggest that this protein is involved in mesodermal formation and neurogenesis during embryonic development. [provided by RefSeq
Sequence: ESRCCRYPLTVDFEAFGWDWIIAPKRYKANYCSGQCEYMFMQKYPHTHLVQQANPRGSAGPCCTPTKMSPINMLYFNDKQQIIYGKIPGMVVDRCGCSSpecifica
| growth differentiation factor 11 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005811 | |
| GDF11 | |
| GDF11 (NP_005802, 310 a.a. ∼ 407 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cytokines | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 3G6 | |
| Mouse monoclonal antibody raised against a partial recombinant GDF11. | |
| GDF11 | |
| BMP-11/BMP11 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 10220 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |