Learn More
growth differentiation factor 3, Mouse, Clone: 1B9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GDF3.
Marca: Abnova H00009573-M02.100ug
Descrizione
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. [provided by RefSeq
Sequence: HRHQLFINFRDLGWHKWIIAPKGFMANYCHGECPFSLTISLNSSNYAFMQALMHAVDPEIPQAVCIPTKLSPISMLYQDNNDNVILRHYEDMVVDECGCGSpecifica
| growth differentiation factor 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_020634 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9573 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA | |
| 1B9 | |
| Mouse monoclonal antibody raised against a partial recombinant GDF3. | |
| GDF3 | |
| GDF3 | |
| GDF3 (NP_065685, 265 a.a. ∼ 364 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cytokines | |
| Primary | |
| Human | |
| Antibody |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.