Learn More
growth differentiation factor 7, Mouse, Clone: 3C2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GDF7.
Marca: Abnova H00151449-M04.100ug
Descrizione
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord. [provided by RefSeq
Sequence: LGWDDWIIAPLDYEAYHCEGLCDFPLRSHLEPTNHAIIQTLLNSMAPDAAPASCCVPARLSPISILYIDAANNVVYKQYEDMVVEACGCRSpecifica
| growth differentiation factor 7 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_182828 | |
| GDF7 | |
| GDF7 (NP_878248, 361 a.a. ∼ 450 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Cytokines | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3C2 | |
| Mouse monoclonal antibody raised against a partial recombinant GDF7. | |
| GDF7 | |
| BMP12 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 151449 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |