Learn More
growth differentiation factor 7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GDF7.
Marca: Abnova H00151449-A01.50uL
Descrizione
This gene encodes a member of the bone morphogenetic protein (BMP) family. BMPs belong to the transforming growth factor-beta superfamily of secreted signalling molecules that regulate diverse processes in growth, repair and embryonic development. In mouse, this gene functions as an inductive signal from the roof plate required for the specification of neuronal identity in the dorsal spinal cord. [provided by RefSeq
Sequence: LGWDDWIIAPLDYEAYHCEGLCDFPLRSHLEPTNHAIIQTLLNSMAPDAAPASCCVPARLSPISILYIDAANNVVYKQYEDMVVEACGCRSpecifica
| growth differentiation factor 7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GDF7. | |
| GDF7 | |
| BMP12 | |
| Mouse | |
| 50 μL | |
| Cytokines | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_182828 | |
| GDF7 | |
| GDF7 (NP_878248, 361 a.a. ∼ 450 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 151449 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.