Learn More
growth differentiation factor 9, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GDF9 protein.
Marca: Abnova H00002661-B01P.50ug
Descrizione
Growth factors synthesized by ovarian somatic cells directly affect oocyte growth and function. Growth differentiation factor-9 (GDF9) is expressed in oocytes and is thought to be required for ovarian folliculogenesis. GDF9 is a member of the transforming growth factor-beta superfamily. [provided by RefSeq
Sequence: MARPNKFLLWFCCFAWLCFPISLGSQASGGEAQIAASAELESGAMPWSLLQHIDERDRAGLLPALFKVLSVGRGGSPRLQPDSRALHYMKKLYKTYATKEGIPKSNRSHLYNTVRLFTPCTRHKQAPGDQVTGILPSVELLFNLDRITTVEHLLKSVLLYNINNSVSFSSAVKCVCNLMIKEPKSSSRTLGRAPYSFTFNSQFEFGKKHKWIQIDVTSLLQPLVASNKRSIHMSINFTCMKDQLEHPSAQNGLFNMTLVSPSLILYLNDTSAQAYHSWYSLHYKRRPSQGPDQERSLSAYPVGEEAAEDGRSSHHRHRRGQETVSSELKKPLGPASFNLSEYFRQFLLPQNECELHDFRLSFSQLKWDNWIVAPHRYNPRYCKGDCPRAVGHRYGSPVHTMVQNIIYEKLDSSVPRPSCVPAKYSPLSVLTIEPDGSIAYKEYEDMIATKCTCRSpecifica
| growth differentiation factor 9 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human GDF9 protein. | |
| GDF9 | |
| GDF9 | |
| GDF9 (NP_005251, 1 a.a. ∼ 454 a.a) full-length human protein. | |
| 50 μg | |
| Cytokines | |
| 2661 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_005260 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.