Learn More
GFAP Mouse, Alexa Fluor 647, Clone: 1B4, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 560298
Descrizione
GFAP (Glial Fibrillary Acidic Protein) is the major protein of glial filaments in differentiated astrocytes. BD Pharmingen offers a panel of monoclonal antibodies (4A11, 1B4, 2E1) that specifically recognize GFAP. They do not cross-react with other intermediate filaments such as vimentin, neurofilament proteins, desmin, keratin, neurotubules or microfilaments.
Host Species: Mouse
Clone: 1B4
Isotype: IgG2b
Species Reactivity: Human
Immunogen: Cow spinal cord homogenate
Intracellular Staining
Specifica
| GFAP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| In vitro Assay | |
| 1B4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Cow spinal cord homogenate | |
| 100 Tests | |
| Cell Biology | |
| Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep | |
| IgG2b |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.