Learn More
GFP Mouse, Alexa Fluor 647, Clone: 1A12-6-18, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 565197
Descrizione
Monoclonal antibody 1A12-6-18 reacts with Green Fluorescent Protein (GFP), which is a 27 kDa bioluminescent protein first purified from the jellyfish, Aequorea victoria . GFP exhibits green fluorescence when exposed to blue or ultraviolet light. Additionally, GFP can be introduced into the genome of and expressed in a wide variety of cell types, making it a useful reporter for tracking gene expression or protein localization. A number of GFP mutants have been developed with varying fluorescence intensity and spectra. Monoclonal antibody 1A12-6-18 is known to react with AcGFP, and is not reactive towards ZsGreen1.
Specifica
| GFP | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide | |
| Affinity Purified | |
| RUO | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1A12-6-18 | |
| Alexa Fluor 647 | |
| Mouse | |
| 50 μg | |
| Primary | |
| IgG1 κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.