Learn More
GPT (M04A), Mouse anti-Human, Clone: M1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GPT.
Marca: Abnova H00002875-M04A.200uL
Descrizione
Glutamate-pyruvate transaminase (EC 2.6.1.2), also known as alanine aminotransferase, catalyzes the reversible conversion of L-alanine and alpha-ketoglutarate to L-glutamate and pyruvate. It has 2 distinct molecular and genetic forms: one cytoplasmic (soluble) (GPT1) and one mitochondrial (GPT2; MIM 138210). See ALTQTL1 (MIM 612363) and ALTQTL2 (MIM 612364) for information on quantitative trait loci influencing the plasma level of alanine aminotransferase.[supplied by OMIM
Sequence: MASSTGDRSQAVRHGLRAKVLTLDGMNPRVRRVEYAVRGPIVQRALELEQELRQGVKKPFTEVIRANIGDAQAMGQRPITFLRQVLALCVNPDLLSSPNFPDDAKKRAERILQACGGHSLGAYSVSSGIQLIREDVARYIERRDGGIPADPNNVFLSTGASDAIVTVLKLLVAGEGHTRTGVLIPIPQYPLYSATLAELGAVQVDYYLDEERAWALDVAELHRALGQARDHCRPRALCVINPGNPTGQVQTRECIEAVIRFAFEERLFLLADEVYQDNVYAAGSQFHSFKKVLMEMGPPYAGQQELASFHSTSKGYMGECGFRGGYVEVVNMDAAVQQQMLKLMSVRLCPPVPGQALLDLVVSPPAPTDPSFAQFQAEKQAVLAELAAKAKLTEQVFNEAPGISCNPVQGAMYSFPRVQLPPRAVERAQELGLAPDMFFCLRLLEETGICVVPGSGFGQREGTYHFRMTILPPLEKLRLLLEELSRFHAKFTLEYSSpecifica
| GPT | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC018207 | |
| GPT | |
| GPT (AAH18207.1, 1 a.a. ∼ 496 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2875 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA | |
| M1 | |
| Mouse monoclonal antibody raised against a full-length recombinant GPT. | |
| GPT | |
| AAT1/ALT1/GPT1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.