Learn More
hydroxyacyl-Coenzyme A dehydrogenase, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human HADH protein.
Marca: Abnova H00003033-D01P.100ug
Descrizione
This gene is a member of the 3-hydroxyacyl-CoA dehydrogenase gene family. The encoded protein functions in the mitochondrial matrix to catalyze the oxidation of straight-chain 3-hydroxyacyl-CoAs as part of the beta-oxidation pathway. Its enzymatic activity is highest with medium-chain-length fatty acids. Mutations in this gene cause one form of familial hyperinsulinemic hypoglycemia. The human genome contains a related pseudogene. [provided by RefSeq
Sequence: MAFVTRQFMRSVSSSSTASASAKKIIVKHVTVIGGGLMGAGIAQVAAATGHTVVLVDQTEDILAKSKKGIEESLRKVAKKKFAENPKAGDEFVEKTLSTIATSTDAASVVHSTDLVVEAIVENLKVKNELFKRLDKFAAEHTIFASNTSSLQITSIANATTRQDRFAGLHFFNPVPVMKLVEVIKTPMTSQKTFESLVDFSKALGKHPVSCKDTPGFIVNRLLVPYLMEAIRLYERGDASKEDIDTAMKLGAGYPMGPFELLDYVGLDTTKFIVDGWHEMDAENPLHQPSPSLNKLVAENKFGKKTGEGFYKYKSpecifica
| hydroxyacyl-Coenzyme A dehydrogenase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human HADH protein. | |
| HADH | |
| HAD/HADH1/HADHSC/HHF4/M/SCHAD/MGC8392/SCHAD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3033 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC000306.2 | |
| HADH | |
| HADH (AAH00306.1, 1 a.a. ∼ 314 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.