missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Helicobacter pylori 19 kDa Monoclonal Antibody (F92B)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA17455
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The MA1-7455 antibody reacts with Helicobacter pylori 19 kDa Ag in bacterial and infected tissue samples. MA1-7455 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7455 antibody was raised against H. pylori 19 kDa antigen.
Helicobacter Pylori (H. Pylori) is known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcimona and low grade lymphoma of mucosa associated lymphoid tissue in the stomach. It is a microaerophilic gram-Negative bacterium, helix-shaped, and with many diverse strains.
Specifica
| Helicobacter pylori 19 kDa | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| F92B | |
| Unconjugated | |
| H. pylori | |
| H. pylori 19 kDa antigen. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto