missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-HMGB1 Ab-2 Matched Antibody Pair, Abnova™
Marca: Abnova H00003146-AP42.1Set
Includes: Capture antibody: mouse monoclonal anti-HMGB1 (100μg); Detection antibody: biotinylated mouse monoclonal anti-HMGB1, IgG1 Kappa (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
Host Species: Mouse, Mouse
Isotype: detection antibody: IgG1, κ
Cross Reactivity: Human
ELISA Pair
Specifica
| HMGB1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human HMGB1. | |
| DKFZp686A04236/HMG1/HMG3/SBP-1 | |
| Mouse | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-HMGB1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-HMGB1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Biotin | |
| HMGB1 | |
| HMGB1 | |
| 1 Set | |
| 3146 | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto