Learn More
IgG2a Rat anti-Human, Invitrogen™
Rat Monoclonal Secondary Antibody
Marca: Invitrogen MA190900
Descrizione
MA1-90900 detects IgG2a from human samples. MA1-90900 has been successfully used in flow cytometry, ELISA, immunohistochemistry (frozen tissue) applications. The MA1-90900 immunogen is: Human lymphocytes Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifica
| Human IgG2a | |
| Monoclonal | |
| 0.1 mg/mL | |
| Rat | |
| Protein G | |
| RUO | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
| LO-DNP-16 | |
| Unconjugated | |
| Human lymphocytes | |
| 100 Tests | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| IgG2a |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.