missing translation for 'onlineSavingsMsg'
Learn More
Learn More
inhibitor of DNA binding 3, dominant negative helix-loop-helix protein, Mouse, Clone: 3E10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ID3.
Marca: Abnova H00003399-M02.100ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Members of the ID family of helix-loop-helix (HLH) proteins lack a basic DNA-binding domain and inhibit transcription through formation of nonfunctional dimers that are incapable of binding to DNA.[supplied by OMIM
Sequence: MKALSPVRGCYEAVCCLSERSLAIARGRGKGPAAEEPLSLLDDMNHCYSRLRELVPGVPRGTQLSQVEILQRVIDYILDLQVVSpecifica
| inhibitor of DNA binding 3, dominant negative helix-loop-helix protein | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002167 | |
| ID3 | |
| ID3 (NP_002158, 1 a.a. ∼ 83 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3E10 | |
| Mouse monoclonal antibody raised against a partial recombinant ID3. | |
| ID3 | |
| HEIR-1/bHLHb25 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 3399 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto