missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IFITM3 Matched Antibody Pair, Abnova
Marca: Abnova H00010410-AP11.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-IFITM3 (100μg); Mouse monoclonal anti-IFITM3, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
ELISA
Specifica
| IFITM3 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IFITM3. | |
| 1-8U/IP15 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-IFITM3 (100 μg) 2. Detection antibody: mouse monoclonal anti-IFITM3, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| IFITM3 | |
| IFITM3 | |
| Affinity Purified | |
| RUO | |
| 10410 | |
| Human | |
| IgG1 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto