Learn More
anti-IgG1, κ, Mouse, PE-Cy™7, Clone: MOPC-21, BD
Marca: BD Biosciences 557872
Descrizione
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Flow Cytometry, Isotype Control
Specifica
| Flow Cytometry | |
| PE-Cyanine7 | |
| Affinity Purified | |
| Undiluted at 4°C |
| MOPC-21 | |
| Mouse | |
| 100 Tests | |
| IgG1 κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.