Learn More
interleukin 28 receptor, alpha (interferon, lambda receptor), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant IL28RA.
Marca: Abnova H00163702-A01.50uL
Descrizione
The protein encoded by this gene belongs to the class II cytokine receptor family. This protein forms a receptor complex with interleukine 10 receptor, beta (IL10RB). The receptor complex has been shown to interact with three closely related cytokines, including interleukin 28A (IL28A), interleukin 28B (IL28B), and interleukin 29 (IL29). The expression of all three cytokines can be induced by viral infection. The cells overexpressing this protein have been found to have enhanced responses to IL28A and IL29, but decreased response to IL28B. Three alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Sequence: QDVTYFVAYQSSPTRRRWREVEECAGTKELLCSMMCLKKQDLYNKFKGRVRTVSPSSKSPWVESEYLDYLFEVEPAPPVLVLTQTEEILSANATYQLPPCSpecifica
| interleukin 28 receptor, alpha (interferon, lambda receptor) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant IL28RA. | |
| IL28RA | |
| CRF2/12/IFNLR/IFNLR1/IL-28R1/LICR2 | |
| Mouse | |
| 50 μL | |
| Membrane Receptors | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_170743 | |
| IL28RA | |
| IL28RA (NP_734464, 51 a.a. ∼ 150 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 163702 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.