Learn More
lamin B1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human LMNB1 protein.
Marca: Abnova H00004001-B01.50uL
Descrizione
The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. This gene encodes one of the two B type proteins, B1. [provided by RefSeq
Sequence: MATATPVPPRMGSRAGGPTTPLSPTRLSRLQEKEELRELNDRLAVYIDKVRSLETENSALQLQVTEREEVRGRELTGLKALYETELADARRALDDTARERAKLQIELGKCKAEHDQLLLNYAKKESDLNGAQIKLREYEAALNSKDAALATALGDKKSLEGDLEDLKDQIAQLEASLAAAKKQLADETLLKVDLENRCQSLTEDLEFRKSMYEEEINETRRKHETRLVEVDSGRQIEYEYKLAQALHEMREQHDAQVRLYKEELEQTYHAKLENARLSSEMNTSTVNSAREELMESRMRIESLSSQLSNLQKESRACLERIQELEDLLAKEKDNSRRMLTDKEREMAEIRDQMQQQLNDYEQLLDVKLALDMEISAYRKLLQGEEERLKLSPSPSSRVTVSRASSSRSVRTTRGKRKRVDVEESEASSSVSISHSASATGNVCIEEIDVDGKFIRLKNTSEQDQPMGGWEMIRKIGDTSVSYKYTSRYVLKAGQTVTIWAANAGVTASPPTDLIWKNQNSWGTGEDVKVILKNSQGEEVAQRSTVFKTTIPEEEEEEEEAAGVVVEEELFHQQGTPRASNRSCAIMSpecifica
| lamin B1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human LMNB1 protein. | |
| LMNB1 | |
| ADLD/LMN/LMN2/LMNB/MGC111419 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 4001 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC012295.1 | |
| LMNB1 | |
| LMNB1 (AAH12295.1, 1 a.a. ∼ 586 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.