missing translation for 'onlineSavingsMsg'
Learn More

LR11 Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

182.97€ - 488.99€

Specifications

Antigene LR11
Clone 48
Concentrazione 250μg/mL
Applicazioni Western Blot
Classificazione Monoclonal
View More Specs

Products 2
Product Code Brand Quantità Price Quantity & Availability  
Product Code Brand Quantità Price Quantity & Availability  
15886039
View Documents
BD Biosciences
611860
50 μg
ONLINE EXCLUSIVE PRICE
183.00€
182.97€ / 1 set
Risparmia 0.03€
0.02% Sconto
View alternative products
Save Up to 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
15896039
View Documents
BD Biosciences
611861
150 μg
ONLINE EXCLUSIVE PRICE
489.00€
488.99€ / 1 set
Risparmia 0.01€
0.00% Sconto
View alternative products
Save Up to 
»
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Description

Description

The low-density lipoprotein receptor (LDLR) functions in lipoprotein transport pathways, and is involved in familial hypercholesterolemia. Homologues of the LDLR may have diverse functions and their ligands may include biological inactive plasma carrier complexes, plasma lipoproteins, yolk precursors, toxins, and extracellular lipoproteins. LR11 (also known as sorLA-1 and gp250) is a lipoprotein receptor homologue that contains 11 LDL receptor ligand binding repeats (LDLRs), 5 LDL receptor “YWTD”repeats, a large fibronectin-type III (FIII) hexarepeat domain similar to neural adhesion proteins, and a domain with similarity to the yeast receptor for vacuolar protein sorting (Vsp10p). LR11mRNA is expressed at high levels in brain, but is also found in liver, pancreas, adrenal gland, and testis. LR11 can bind the ER and Golgi localized receptor associated protein (RAP), which binds to many members of the LDLR family and prevents aggregation of ligands to the LDLRs. In addition, LR11 binds apolipoprotein E-containing lipoproteins. Thus, LR11 is a lipoprotein binding receptor that may have additional functions related to protein sorting and neuronal development.

Host Species: Mouse
Clone: 48
Isotype: IgG2a
Species Reactivity [for Features Main]: Rat
Immunogen: Human LR11 aa. 1220-1337

Immunofluorescence, Western Blotting

Specifications

Specifications

LR11
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
SorLA; gp250; Lipoprotein Receptor-11
48
Western Blot
Unconjugated
RUO
Human, Mouse, Rat
Human LR11 aa. 1220-1337
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.