missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-POGK Matched Antibody Pair, Abnova™
Marca: Abnova H00057645-AP21.1Set
Includes: Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-POGK (100μg); Detection antibody: mouse polyclonal anti-POGK (40μL); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human POGK.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
ELISA Pair (Recombinant Protein)
Specifica
| POGK | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human POGK. | |
| BASS2/KIAA1513/KIAA15131/LST003 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-POGK (100 μg) 2. Detection antibody: mouse polyclonal anti-POGK (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| POGK | |
| POGK | |
| Affinity Purified | |
| RUO | |
| 57645 | |
| Human |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto