Learn More
pogo transposable element with KRAB domain, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human POGK protein.
Marca: Abnova H00057645-D01P.100ug
Descrizione
The exact function of the protein encoded by this gene is not known. However, this gene product contains a KRAB domain (which is involved in protein-protein interactions) at the N-terminus, and a transposase domain at the C-terminus, suggesting that it may belong to the family of DNA-mediated transposons in human. [provided by RefSeq
Sequence: MESTAYPLNLSLKEEEEEEEIQSRELEDGPADMQKVRICSEGGWVPALFDEVAIYFSDEEWEVLTEQQKALYREVMRMNYETVLSLEFPFPKPDMITRLEGEEESQNSDEWQLQGGTSAENEESDVKPPDWPNPMNATSQFPQPQHFDSFGLRLPRDITELPEWSEGYPFYMAMGFPGYDLSADDIAGKFQFSRGMRRSYDAGFKLMVVEYAESTNNCQAAKQFGVLEKNVRDWRKVKPQLQNAHAMRRAFRGPKNGRFALVDQRVAEYVRYMQAKGDPITREAMQLKALEIAQEMNIPEKGFKASLGWCRRMMRRYDLSLRHKVPVPQHLPEDLTEKLVTYQRSVLALRRAHDYEVAQMGNADETPICLEVPSRVTVDNQGEKPVLVKTPGREKLKITAMLGVLADGRKLPPYIILRGTYIPPGKFPSGMEIRCHRYGWMTEDLMQDWLEVVWRRRTGAVPKQRGMLILNGFRGHATDSVKNSMESMNTDMVIIPGGLTSQLQVLDVVVYKPLNDSVRAQYSNWLLAGNLALSPTGNAKKPPLGLFLEWVMVAWNSISSESIVQGFKKCHISSNLEEEDDVLWEIESELPGGGEPPKDCDTESMAESNSpecifica
| pogo transposable element with KRAB domain | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human POGK protein. | |
| POGK | |
| BASS2/KIAA1513/KIAA15131/LST003 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_017542 | |
| POGK | |
| POGK (NP_060012.3, 1 a.a. ∼ 609 a.a) full-length human protein. | |
| 100 μg | |
| Apoptosis | |
| 57645 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.