Learn More
PPAR gamma/NR1C3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Marca: Novus Biologicals NBP2-22106SS
Descrizione
PPAR gamma/NR1C3 Polyclonal specifically detects PPAR gamma/NR1C3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifica
| PPAR gamma/NR1C3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CIMT1, NR1C3GLM1, Nuclear receptor subfamily 1 group C member 3, peroxisome proliferator-activated receptor gamma, peroxisome proliferator-activated receptor gamma 1, PPAR gamma, PPARG1peroxisome proliferative activated receptor, gamma, PPARG2PPARgamma, PPAR-gamma | |
| Rabbit | |
| 55 kDa | |
| 0.025 mL | |
| Primary | |
| Immunogen has 87% homology to rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Immunohistochemistry 1:100, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100 | |
| P37231 | |
| PPARG | |
| A synthetic peptide made to an internal portion of the human PPAR gamma protein (between residues 20-120) [UniProt P37231] | |
| Affinity Purified | |
| RUO | |
| 5468 | |
| Human, Mouse, Rat | |
| IgG |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
For Research Use Only