Learn More
anti-S100A8 Matched Antibody Pair, Ab-2, Abnova
Marca: Abnova H00006279-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-S100A8 (100μg); Mouse polyclonal anti-S100A8 (40μL); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Descrizione
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and as a cytokine. Altered expression of this protein is associated with the disease cystic fibrosis.
ELISA
Specifica
| S100A8 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human S100A8. | |
| 60B8AG/CAGA/CFAG/CGLA/CP-10/L1Ag/MA387/MIF/MRP8/NIF/P8 | |
| Mouse, Rabbit | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-S100A8 (100 μg) 2. Detection antibody: mouse polyclonal anti-S100A8 (40 ul) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
| ELISA | |
| Unconjugated | |
| S100A8 | |
| S100A8 | |
| Affinity Purified | |
| RUO | |
| 6279 | |
| Human | |
| Serum |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.