missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A8, Human, Matched Antibody Pair (AP52), Abnova™
Marca: Abnova H00006279-AP52.1Set
Includes: Capture antibody: mouse monoclonal anti-S100A8 (100μg); Detection antibody: rabbit purified polyclonal anti-S100A8 (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human S100A8.
Host Species: Mouse, Rabbit
Species Reactivity: Hu
ELISA Pair (Transfected Lysate)
Specifications
| S100A8 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human S100A8. | |
| 60B8AG/CAGA/CFAG/CGLA/CP-10/L1Ag/MA387/MIF/MRP8/NIF/P8 | |
| Mouse, Rabbit | |
| RUO | |
| 6279 | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| S100A8 | |
| S100A8 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-S100A8 (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-S100A8 (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction