Learn More
anti-S100A9, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marca:  Abnova H00006280-PW2.1Set
Codice prodotto. 16112604
  • 843.00€ / 1 set
Includes: Antibody pair for IP: mouse monoclonal anti-S100A9 (300μg), Antibody pair for WB: rabbit purified polyclonal anti-S100A9 (50ug)
Questo articolo non è restituibile. Consulta la politica di reso
Specifica
S100A9
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
60B8AG/CAGB/CFAG/CGLB/L1AG/LIAG/MAC387/MIF/MRP14/NIF/P14
Mouse, Rabbit
RUO
6280
Human
Liquid
Immunoprecipitation, Western Blot
Unconjugated
S100A9
S100A9
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: mouse monoclonal anti-S100A9 (300 ug) 2. Antibody pair for WB: rabbit purified polyclonal anti-S100A9 (50ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Certificati
Promozioni
Promozioni
For Research Use Only