Learn More
SARS Nucleocapsid Protein Rabbit anti-SARS-CoV-2, SARS-CoV, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 49 publications
Marca: Novus Biologicals NB100-56576
Descrizione
SARS Nucleocapsid Protein Polyclonal specifically detects SARS Nucleocapsid Protein in SARS-CoV-2, SARS-CoV, Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC.Specifica
| SARS Nucleocapsid Protein | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, N, N protein, N structural protein, NC, Nucleocapsid protein, Nucleoprotein, Protein N, SARS coronavirus N protein, SARS coronavirus nucleocapsid protein, SARS CoV, SARS CoV N protein, SARS CoV nucleocapsid protein, SARS N protein, SARS Nucleoprotein, SARSCoV, SARSCoV N protein, SARSCoV nucleocapsid protein, SARS-CoV-2 N protein, Severe acute respiratory syndrome | |
| The antibody was developed by immunizing Rabbit with a synthetic peptide corresponding to amino acids 399-411 (DLDDFSKQLQQSM-C) from the N (SARS Nucleocapsid) for the Human SARS coronavirus (Genbank accession no. YP_009724397.2) | |
| 0.1 mg | |
| Primary | |
| SARS-CoV-2, SARS-CoV, Virus | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), In Situ Hybridization (ISH) | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin, Dual RNAscope ISH-IHC | |
| NP_006504.2 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| SARS Nucleocapsid protein. Dot Blot results using recombinant proteins for cross-reactivity testing revealed high reactivity to SARS-CoV-2 Nucleocapsid protein (NBP2-90975) and low/no reactivity towards H1N1 (NBP1-99041). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Il programma Encompass di Fisher Scientific offre articoli che non fanno parte del nostro portafoglio di distribuzione. Questi prodotti in genere non hanno immagini o descrizioni dettagliate. Tuttavia, ci impegniamo a migliorare la vostra esperienza di acquisto. Si prega di utilizzare il modulo sottostante per fornire un feedback relativo al contenuto di questo prodotto.
For Research Use Only