Learn More
3M™ Anti-Scratch/Anti-Fog Safety Glasses
Click to view available options
Lens Tint:
Amber
Gray
Shade 5
Trasparente
Dimensione della confezione:
Pezzo
pezzi 20
Descrizione
- Very lightweight, sleek and modern design safety glasses offering excellent coverage and field of vision
- Strong polycarbonate lens for impact resistance
- Optical class 1 for maximum clarity
- Inclinable lens for easy adjustment and maximum comfort
- Soft, flexible temples for wearer comfort
- Anti-Scratch and Anti-Fog coating for increased durability and improved vision
- ™™
- Safety glasses designed with user comfort and convenience in mind
- Strong polycarbonate lens for impact resistance
- Lightweight (26g) for maximum comfort all day
- Integrated browguard for increased protection
- Soft inner padding on temple for increased comfort and stability when worn
- Adjustable temple lengths in three positions for optimal comfort and fit
- Inclinable lens for easy adjustment and maximum comfort
Specifica
Specifica
| Certificazione/Conformità | CE Approved, EN 166:2001 |
| Lens Coating | Antiappannamento, Resistente ai graffi |
| Lens Tint | Trasparente |
| Product Type | Safety glasses |
| Colore telaio | Grigio scuro |
| Lens Material | Policarbonato |