Learn More
Sec8 Mouse, Unlabeled, Clone: 14, BD
Mouse Monoclonal Antibody
377.71€
Specifica
| Antigene | Sec8 |
|---|---|
| Clone | 14 |
| Concentrazione | 250μg/mL |
| Applicazioni | Western Blot |
| Classificazione | Monoclonal |
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|---|---|---|---|---|---|---|---|---|---|
| Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
|
15845569
|
BD Biosciences
610658 |
50 μg |
378.00€ 377.71€ / 1 set Risparmia 0.29€ 0.08% Sconto |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Signal transmission between neurons is regulated by the release of neurotransmitters at the synapse. This process, which is controlled by a complex pathway of membrane trafficking in the presynaptic nerve terminal, leads to membrane fusion and neurotransmitter secretion. Sec8 is a hydrophilic protein of 975 amino acids that is highly expressed in brain and kidney. It is homologus with the yeastsecretory protein, Sec8p. In yeast, Sec8 is essential for the Golgi-to-plasma membrane traffic of proteins during constitutive secretion. In rat brain, Sec8 colocalizes with Rab3 and Syntaxin-1a, two important proteins that regulate the fusion of the synaptic vesicle to the plasma membrane. The primary structure of Sec8, a coiled-coil domain (residues 34-99), is commonly found in other proteins important in secretory pathways.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifica
| Sec8 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG2b | |
| Affinity Purified |
| 14 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Chicken, Human, Mouse, Rat | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.