missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-T, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Marca: Abnova H00006862-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-T, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-T, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
| T | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human T. | |
| MGC104817/TFT | |
| Mouse | |
| RUO | |
| 6862 | |
| Human | |
| Liquid |
| ELISA | |
| Biotin | |
| T | |
| T | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-T, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-T, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction