Learn More
TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human TAF1C protein.
Marca: Abnova H00009013-D01P.100ug
Descrizione
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified. [provided by RefSeq
Sequence: MLPPLIDPWDPGLTARDLLFRGGYRYRKRPRVVLDVTEQISRFLLDHGDVAFAPLGKLMLENFKLEGAGSRTKKKTVVSVKKLLQDLGGHQPWGCPWAYLSNRQRRFSILGGPILGTSVASHLAELLHEELVLRWEQLLLDEACTGGALAWVPGRTPQFGQLVYPAGGAQDRLHFQEVVLTPGDNPQFLGKPGRIQLQGPVRQVVTCTVQGETLLAVRSDYHCAVWKFGKQWQPTLLQAMQVEKGATGISLSPHLPGELAICSRSGAVCLWSPEDGLRQIYRDPETLVFRDSSSWRWADFTAHPRVLTVGDRTGVKMLDTQGPPGCGLLLFRLGAEASCQKGERVLLTQYLGHSSPKCLPPTLHLVCTQFSLYLVDERLPLVPMLKWNHGLPSPLLLARLLPPPRPSCVQPLLLGGQGGQLQLLHLAEGASVPRLAGPPQSLPSRIDSLPAFPLLEPKIQWRLQERLKAPTIGLAAVVPPMPSAPTPGLVLFQLSAAGDVFYQQLRPQVDSSLRRDAGPPGDTQPDCHAPTASWTSQDTAGCSQWLKALLKVPLAPPVWTAPTFTHRQMLGSTELRREEEEGQRLGVLRKAMARGQLLLQRDLGSLPAAEPPPAPESGLEDKLSERLGEAWAGRGAAWWERQQGRTSEPGRQTRRPKRRTQLSSSFSLSGHVDPSEDTSSPHSPEWPPADALPLPPTTPPSQELTPDACAQGVPSEQRQMLRDYMAKLPPQRDTPGCATTPPHSQASSVRATRSQQHTPVLSSSQPLRKKPRMGFSpecifica
| TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human TAF1C protein. | |
| TAF1C | |
| MGC:39976/SL1/TAFI110/TAFI95 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC028131.1 | |
| TAF1C | |
| TAF1C (AAH28131.1, 1 a.a. ∼ 775 a.a) full-length human protein. | |
| 100 μg | |
| Transcription Regulation | |
| 9013 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.