missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TK1, Human, Matched Antibody Pair (AP42), Abnova™
To detect and quantify protein level of human
Marca: Abnova H00007083-AP42.1Set
Includes: Capture antibody: mouse monoclonal anti-TK1, IgG2a Kappa (100μg) , Detection antibody: alkaline phosphatase (ALP) conjugated mouse monoclonal anti-TK1 (100μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifica
| TK1 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TK1. | |
| TK2 | |
| Mouse | |
| RUO | |
| 7083 | |
| Human | |
| Liquid |
| ELISA | |
| Alkaline Phosphatase (AP) | |
| TK1 | |
| TK1 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TK1, IgG2a Kappa (100 ug) 2. Detection antibody: alkaline phosphatase (ALP) conjugated mouse monoclonal anti-TK1, Non isotype (100 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
| Store mouse anti-TK1 monoclonal antibody at -20°C or lower. Store alkaline phosphatase (ALP) conjugated mouse anti-TK1 monoclonal antibody at 4 °C Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20 °C or 4 |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto