Learn More
Antibacteria Mats
Codice prodotto. p-8010417
Click to view available options
Colore:
Blue
White
Dimensioni (L x P):
61 x 91 cm
91 x 114 cm
91 x 114 cm
Dimensione della confezione:
pezzi 120
Codice prodotto. 11886383
Marca: CEEC Control Environ Eqpt 177548
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Use CEEC Control Environ Eqpt Corp™ Antibacteria Mats to bond directly to the floor or in aluminum. After use, detach the sheets, which are made of polyethylene.
  • Eliminate bacterial contamination
  • Sheets numbered 30 to 1
  • Supplied in cartons of 4 stacks of 30 sheets polyethylene
Specifications
Dimensioni (L x P) 61 x 91 cm
Quantità 120/Pk.
Colore White
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.