missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANTXR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15091-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colorectal cancer. The encoded protein has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax. The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to this receptor protein mediates delivery of toxin components to the cytosol of cells. Once inside the cell, the other two components of anthrax toxin, edema factor (EF) and lethal factor (LF) disrupt normal cellular processes. Three alternatively spliced variants that encode different protein isoforms have been described.Specifica
| ANTXR1 | |
| Polyclonal | |
| Unconjugated | |
| ANTXR1 | |
| anthrax toxin receptor 1, ANTXR1, ATR, TEM8, Tumor endothelial marker 8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 362393, 69538, 84168 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q0PMD2, Q9CZ52, Q9H6X2 | |
| Antxr1 | |
| ANTXR1 Fusion Protein Ag4626 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto