Learn More
Tocris Bioscience™ Aprotinin
Click to view available options
:
10mg
Descrizione
Aprotinin is a competitive serine protease inhibitor. Reversibly binds to and blocks the enzymatic active site. Inhibits a range of serine proteases including trypsin, chymotrypsin, kallikrein and plasmin. Inhibits cytopathogenic effect of SARS-CoV-2 and double-stranded RNA formation in SARS-CoV-2-infected cells.
This product is from Bovine Lung.
Specifica
Specifica
| Peso molecolare | 6511.48 |
| Product Type | Aprotinin |
| Quantità | 10 mg |
For Research Use Only