Learn More
3M™ Nomad™ Aqua Textile Drop Down Mat, Series 85
Click to view available options
Dimensioni (L x P):
150 x 91.4 cm
200 x 130 cm
300 x 130 cm
300 x 200 cm
600 x 130 cm
Colore:
Black
Gray
Dimensione della confezione:
Pezzo
Descrizione
- High performing textile entrance mat, with advanced water removal and retention properties
- Stops 80% of dirt and moisture at the door
- Entrance mat for high traffic applications (1500 to 5000 crossings per day)
- Phthalate-free, slip-resistant PVC backing
- Excellent appearance retention
- Long lasting and easy to clean
- Available as drop down mats or in rolls
Specifica
Specifica
| Dimensioni (L x P) | 200 x 130 cm |
| Da utilizzare con (applicazione) | Heavy traffic areas |
| Modello | Nomad™ Aqua, Series 85 |
| Resistenza | Slip-resistant |
| Spessore (sistema metrico) | 7.5 mm |
| Colore | Gray |
| Lunghezza (sistema metrico) | 200 cm |
| Materiale | PVC (Backing) |
| Larghezza (sistema metrico) | 130 cm |