Learn More
Chemsplash AquaRepel Non Toxic, Water Repellant Coverall
GREENER_CHOICE
Product Code. 30058157
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
pezzi 50
This item is not returnable. View return policy
Product Code. 30058157
Brand: Chemsplash 2619.300.009
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
The Aqua Repel Coverall is made from 40 gsm Polypropylene coated fabric. Is water repellent and provides protection against non-hazardous limited liquid splash.
Chemsplash Aqua Repel Coverall
  • 40 GSM Polypropylene Coated Fabric
  • Non Toxic, Waterproof Fabric
  • Hood With Elasticated Band
  • Elasticated Cuffs and Ankles
  • Half Elasticated Waist
  • Zipped Front With Adhesive Zip Flap
  • PFAS Free
Description: The Chemsplash Aqua Repel Coverall is made from 40 GSM Polypropylene Coated Fabric. This is a Cat 1 suit which is lightweight and water repellent, providing protection against non-hazardous limited liquid splash.
Specifications
Product Type Hooded Coveralls
Size 3X-Large
Sesso Unisex
Materiale Polypropylene
Colore Blue
Monouso Single-use
Stile vita Elastic
Ankle Style Elastic
Stile polso Elastic
Stile collo Hooded
Hood Type Attached
Tipo di chiusura Ties
Tipo di giuntura Stitched
Sterilità Non-sterile
Resistenza Liquid
Senza lattice Yes
N. di tasche None
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.