Learn More
ARF5 Mouse anti-Human, Unconjugated, Clone: 2B3B1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67279-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The ARF5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARF5. This antibody recognizes human, mouse, rat, pig antigen. The ARF5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
| ARF5 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 2B3B1 | |
| Unconjugated | |
| 150 μL | |
| 381 | |
| IgG1 |