Learn More
Abnova™ ARHGEF5 Recombinant Protein
Descrizione
Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization.
- Theoretical MW (kDa): 82.83
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MGGFSRRCSKLINSSQLLYQEYSDVVLNKEIQSQQRLESLSETPGPSSPRQPRKALVSSESYLQRLSMASSGSLWQEIPVVRNSTVLLSMTHEDQKLQEVKFELIVSEASYLRSLNIAVDHFQLSTSLRATLSNQEHQWLFSRLQDVRDVSATFLSDLEENFENNIFSFQVCDVVLNHAPDFRRVYLPYV
TNQTYQERTFQSLMNSNSNFREVLEKLESDPVCQRLSLKSFLILPFQRITRLKLLLQNILKRTQPGSSEEAEATKAHHALEQLIRDCNNNVQSMRRTEELIYLSQKIEFECKIFPLISQSRWLVKSGELTALEFSASPGLRRKLNTRPVHLHLFNDCLLLSRPREGSRFLVFDHAPFSSIRGEKCEMKLHGPHKNLFRLFLRQNTQGAQAEFLFRTETQSEKLRWISALAMPREELDLLECYNSPQVQCLRAYKPRENDELALEKADVVMVTQQSSDGWLEGVRLSDGERGWFPVQQVEFISNPEVRAQNLKEAHRVKTAKLQLVEQQA
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifica
Specifica
| Numero di accesso | AAH14555 |
| Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulazione | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| ID gene (immissione) | 7984 |
| Peso molecolare | 82.83 |
| Nome | ARHGEF5 (Human) Recombinant Protein (P01) |
| Intervallo di pH | 8 |
| Metodo di preparazione | In vitro wheat germ expression system |
| Metodo di purificazione | Glutathione Sepharose 4 Fast Flow |
| Analisi di controllo qualità | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.