Learn More
Skytec Aria 360 Nitrile Foam Gloves
GREENER_CHOICE
Codice prodotto. p-7250636
Click to view available options
Size:
6, X-Small
7, Small
8, Medium
9, Large
10, X-Large
11, 2X-Large
Dimensione della confezione:
10 paia
Codice prodotto. 17851654
Marca: Skytec SKG00022FB
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Aria 360 Gloves are lightweight nitrile foam gloves with an outstanding durability and grip, eco friendly liner and touch screen capabilities.
  • GeniumTM is an eco friendly liner with 1/3 of the ingredients by weight that are sustainably sourced and that requires 30% less energy to be produced
  • Touchscreen capability on thumb and index finger
  • Anatomically designed with moisture wicking to deliver great comfort
  • Excellent grip in all conditions (dry wet, oily)
  • Low particle shedding for sensitive applications
  • Super soft liner
  • Patented nitrile foam palm coating enhances protection
  • Sanitized™ antimicrobial finish provides extra freshness
  • Light heat protection up to 100°C for a short period
Specifica
ANSI Abrasion Rating ANSI 3
Certificazione/Conformità ANSI Abrasion, ANSI Puncture
Certifications/Compliance (Europe) EN388, EN420, EN 407
Chlorinated No
Area rivestita Palm
Struttura Knit
Coating Material Nitrile Foam
Colore Gray, Black
Cut Resistance Rating ANSI A1
Finitura esterna Partially Textured
Hand Specific
Latex Free No
Materiale Nylon, Spandex, Nitrile
Product Type Touchcreen Work Glove
Size 6, X-Small
Da utilizzare con (applicazione) General handling
Stile polso guanto Elasticated
Valori nominali perforazione ANSI/ISEA 105-16 Level 2
Resistente a Abrasion, Punctures
Stile pollice Straight
Lunghezza (sistema imperiale) 9 in.
Lunghezza (sistema metrico) 240 mm