Learn More
Novus Biologicals™ ARNT/HIF-1 beta Antibody Pack
For Research applications
Marca: Novus Biologicals™ NB100-982
Includes:
- 1.NB100-110: ARNT/HIF-1 beta Antibody
- 2.NB100-124: ARNT/HIF-1 beta Antibody (H1β234)
Descrizione
Specifica
| ARNT/HIF-1 beta | |
| Unconjugated | |
| ARNT protein, aryl hydrocarbon receptor nuclear translocator, BHLHE2, HIF-1 Beta, HIF1B, HIF-1-beta, HIF1-beta, HIF1BETA, hypoxia-inducible factor 1, beta subunit, Hypoxia-inducible factor 1-beta, nuclear translocator | |
| Mouse | |
| 2 Vials | |
| 405 | |
| 1.NB100-110: ARNT/HIF-1 beta Antibody 2.NB100-124: ARNT/HIF-1 beta Antibody (H1β234) |
|
| Store at 4C. Do not freeze. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoprecipitation | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunoblotting, Chromatin Immunoprecipitation (ChIP) | |
| ARNT | |
| See individual datasheets. | |
| Cancer, Hypoxia | |
| See individual datasheets. | |
| Human, Mouse, Rat, Bovine, Ferret, Sheep |