Learn More
Abeba™ Arrow 38200 Shoes
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
Dimensione della confezione:
1 paio
Descrizione
- CE EN ISO 20347:2012 OB A, FO SRA
- Smooth leather white
- Slip resistance PU/PU outsole sole
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap foldable, adjustable
- Microfiber heel patch
- Lining microfiber, respiration absorbing
Specifica
Specifica
| Certificazione/Conformità | CE EN ISO 20347:2012 OB A, FO SRA |
| Colore | White |
| Materiale rivestimento interno | Microfiber |
| Materiale | Leather |
| Size | 47 |
| Caratteristiche suola | Slip-resistant |
| Materiale suola | Polyurethane |
| Stile | 38200 |