missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16645-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.Specifica
| ASL | |
| Polyclonal | |
| Unconjugated | |
| ASL | |
| argininosuccinate lyase, Arginosuccinase, ASAL, ASL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109900, 435, 59085 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P04424, P20673, Q91YI0 | |
| Asl | |
| ASL Fusion Protein Ag10007 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto