Learn More
Cole-Parmer™ Aspirator Pump
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Portata:
0.63 CFM
Dimensione della confezione:
Pezzo
Descrizione
- Produce pulseless vacuums up to 29.5 in.
- Built-in circulating pump and water tank makethese units portable and eliminate water waste
- Simple to operate. Just fill the tank and turn on the pump
- Circulating pump continuously forces water across a set of venturies. Operation is quiet, clean and efficient.
Specifica
Specifica
| Vuoto max | 29.5 Hg |
| Amperaggio | 0.35 amp |
| Materiale | Nickel-Coated Brass, Polypropylene, Silicone, 304 Stainless Steel |
| Portata | 0.63 CFM |
| Tipo di pompa | Vacuum System |
| Tensione | 230 V |