Learn More
Abnova™ ATP6V1B2 (Human) Recombinant Protein
Human ATP6V1B2 full-length ORF ( AAH03100, 1 a.a. - 511 a.a.) recombinant protein with GST-tag at N-terminal.
Marca:  Abnova™ H00000526-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16172071
  • 528.00€ / 25µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Sequence: MALRAMRGIVNGAAPELPVPTGGPAVGAREQALAVSRNYLSQPRLTYKTVSGVNGPLVILDHVKFPRYAEIVHLTLPDGTKRSGQVLEVSGSKAVVQVFEGTSGIDAKKTSCEFTGDILRTPVSEDMLGRVFNGSGKPIDRGPVVLAEDFLDIMGQPINPQCRIYPEEMIQTGISAIDGMNSIARGQKIPIFSAAGLPHNEIAAQICRQAGLVKKSKDVVDYSEENFAIVFAAMGVNMETARFFKSDFEENGSMDNVCLFLNLANDPTIERIITPRLALTTAEFLAYQCEKHVLVILTDMSSYAEALREVSAAREEVPGRRGFPGYMYTDLATIYERAGRVEGRNGSITQIPILTMPNDDITHPIPDLTGYITEGQIYVDRQLHNRQIYPPINVLPSLSRLMKSAIGEGMTRKDHADVSNQLYACYAIGKDVQAMKAVVGEEALTSDDLLYLEFLQKFERNFIAQGPYENRTVFETLDIGWQLLRIFPKEMLKRIPQSTLSEFYPRDSAKH
Specifica
Specifica
AAH03100
ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B2
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6V1B2
GST
526
Wheat germ expression system
25 μg
ATP6B1B2, ATP6B2, HO57, VATB, VPP3, Vma2
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer