missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP6V1B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15097-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
ATP6V1B2 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. ATP6V1B2 is one of two V1 domain B subunit isoforms and is the only B isoform highly expressed in osteoclasts.Specifica
| ATP6V1B2 | |
| Polyclonal | |
| Unconjugated | |
| ATP6V1B2 | |
| ATP6B1B2, ATP6B2, ATP6V1B2, HO57, V ATPase subunit B 2, VATB, Vma2, VPP3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117596, 11966, 526 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21281, P62814, P62815 | |
| ATP6V1B2 | |
| ATP6V1B2 Fusion Protein Ag7174 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto