missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Atrial Natriuretic Peptide (ANP) (1-28), rat TFA
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
Specifications
Specifications
| Purezza | 98.74% |
| Content And Storage | Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Da utilizzare con (applicazione) | COVID-19-immunoregulation |
| Peso molecolare | 3176.43 g/mol |
| Product Type | Peptides |
| Quantità | 1 mg |
| Shipping Condition | Room temperature |
| Sinonimo | Atrial natriuretic factor (1-28) (rat) TFA |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction