missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATXN1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA549084
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Predicted to react with mouse based on sequence homology.
Chromatin-binding factor that repress Notch signaling in the absence of Notch intracellular domain by acting as a CBF1 corepressor. Binds to the HEY promoter and might assist, along with NCOR2, RBPJ-mediated repression (PubMed:21475249). Can suppress ATXN1 cytotoxicity in spinocerebellar ataxia type 1 (SCA1). In concert with CIC and ATXN1, involved in brain development. [UniProt].
Specifica
| ATXN1L | |
| Polyclonal | |
| Unconjugated | |
| ATXN1L | |
| ankyrin repeat and SOCS box containing 16; ankyrin repeat and SOCS box protein 16; ankyrin repeat and SOCS box-containing 16; ankyrin repeat and SOCS box-containing protein 16; ASB16; ASB-16; ataxin 1 like; ataxin 1-like; ataxin-1-like; ATXN1L; BOAT; BOAT1; Brother of ataxin-1; brother of ATXN1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 342371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P0C7T5 | |
| ATXN1L | |
| KLH conjugated synthetic peptide between 385-411 amino acids from the Central region of human ATXN1L. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto