Learn More
3M™ Respiratori per particolato
Codice prodotto. 12014128
Click to view available options
:
Filter efficiency: FFP1; Unvalved; Shape: cup
Codice prodotto. 12014128
Marca: 3M™ 7000034739
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
Fits a wide range of face shapes and sizes
  • Low breathing resistance filter technology for easier breathing
  • Ingenious three-panel design accommodates greater facial movement during speech for more comfortable wear
  • An embossed top panel to reduce fogging of eyewear
  • A sculpted nose panel for improved compatibility with eyewear
  • An innovative chin tab for ease of donning and adjustment of the respirator on the face
  • Supplied in hygienic packaging to help protect the respirator from contamination before use, also allows practical storage and dispensing in the workplace.
  • Cool Flow™ valve reduces heat build-up to offer workers comfortable protection - even in hot and humid conditions