Learn More
Stainless Steel, Austenitic, ARMI∣MBH
Codice prodotto. 17900426
Click to view available options
Dimensione della confezione:
Pezzo
Codice prodotto. 17900426
Marca: ARMI/MBH MBH13X 12534 X
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Stainless Steel, Austenitic, Ø40x20mm
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Tungsten/Phosphorus/Tantalum/Aluminium/Copper/Cobalt/Carbon/Vanadium/Niobium/Titanium/Manganese/Silicon/Molybdenum/Nickel/Chromium
CAS 7439-89-6
Forma Disc
Concentration or Composition (by Analyte or Components) Tungsten 0.01 %/Phosphorus 0.0192 %/Tantalum 0.031 %/Aluminium 0.0485 %/Copper 0.0586 %/Cobalt 0.0602 %/Carbon 0.0716 %/Vanadium 0.11 %/Niobium 0.201 %/Titanium 0.348 %/Manganese 0.589 %/Silicon 0.811 %/Molybdenum 2.04 %/Nickel 8.5 %/Chromium 17.71 %