Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
Sfoglia Tutto MBP Molecular BioProducts Prodotti
Click to view available options
Volume (sistema metrico):
20 μL
130 µl
200 μL
Filtrato:
Con filtro
Non-filtered
Sterilità:
Non sterile
Sterile
Sí
Fit for Purpose:
Certified Quality
Sterile
Sterile and PCR Ready
Dimensione della confezione:
pezzi 4800
Descrizione
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
- Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
- Lot-tested during production on actual workstation for which they are created
- Extended length tips compatible with 96-, 384-, and 1536-deep well plates
- Low percentage coefficient of variation (%CV)
Specifica
Specifica
| Volume (sistema metrico) | 20 μL |
| Filtrato | Non-filtered |
| Sterilità | Non sterile |
| Colore | Trasparente |
| Formato | In rack |
| Stile punta | Extended Length |
| Da utilizzare con (apparecchiatura) | Beckman Coulter Biomek 1000/2000 |
| Product Line | Beckman |
| Fit for Purpose | Certified Quality |