Learn More
Thermo Scientific™ Automation Tips for BioTek™ Pipetting Systems
Click to view available options
Volume (sistema metrico):
50 μL
80 μL
100 μL
150 μL
200 μL
Sterilità:
Non sterile
Sterile
Filtrato:
Con filtro
Non-filtered
Dimensione della confezione:
pezzi 4800
Descrizione
Increase pipetting performance with Thermo Scientific™ Automation Tips forBioTek™ Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: BioTek Precision and Precision XS.
- Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifica
Specifica
| Volume (sistema metrico) | 100 μL |
| Filtrato | Non-filtered |
| Sterilità | Non sterile |
| Colore | Trasparente |
| Formato | In rack |
| Stile punta | Low Retention |
| Da utilizzare con (apparecchiatura) | BioTek Pipetting System |
| Product Line | BioTek |